Would you like to log in again?
Castroviejo Needle Holder, Straight Lock Extra Delicate, Roboz
Supplier: Roboz
RS-6416
100189-258EA 535.85 USD InStock
100189-258
Castroviejo Needle Holder, Straight Lock Extra Delicate, Roboz
Forceps
Castroviejo Needle Holder, 0.6 mm wide jaws to hold 7-0 to 10-0 sutures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty