Would you like to log in again?
Metzenbaum (Lahey) Scissors 7" Curved Tungsten Carbide Sure Cut Extra Delicate, Roboz Surgical
Supplier: Roboz
RS-6955SC
100189-264EA 175.85 USD InStock
100189-264
Metzenbaum (Lahey) Scissors 7" Curved Tungsten Carbide Sure Cut Extra Delicate, Roboz Surgical
Scissors
Metzenbaum (Lahey) Scissors; Curved; Extra Delicate; Sure Cut Carbide Blades; Blade Length 35 mm; 7" Overall Length.
These Tungsten Carbide reinforced Sure Cut scissors make cutting easy and will last over 4 times longer than stainless steel scissors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty