Would you like to log in again?
Knapp Scissors 4" Straight Blunt/Blunt, Roboz Surgical
Supplier: Roboz
RS-5962
100189-300EA 145.38 USD InStock
100189-300
Knapp Scissors 4" Straight Blunt/Blunt, Roboz Surgical
Scissors
Knapp Scissors; Straight; Blunt-Blunt; 27 mm Blade Length; 4" Overall Length
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty