Would you like to log in again?
Micro Injection Needle, 32 Gauge 45 Degree, Roboz Surgical
Supplier: Roboz
IN-905
100189-334EA 114.92 USD InStock
100189-334
Micro Injection Needle, 32 Gauge 45 Degree, Roboz Surgical
Syringe Needles
Micro Injection Needle; 45 Degree Angle; .009" Outside Diameter, .003" Inside Diameter; 32 Gauge; 10 mm Needle Length.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty