Would you like to log in again?
Tube Occluding Forceps, 5.75", Serrated Straight, Roboz
Supplier: Roboz
RS-7476
100189-380EA 119.08 USD InStock
100189-380
Tube Occluding Forceps, 5.75", Serrated Straight, Roboz
Forceps
St
Vincent Tube Occluding Forceps; Narrow Jaws; Straight, Serrated; Lockguard; 5.75" Length. The jaw width is 4 mm at 5 mm from the tip
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty