Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fine Points, 3.5" (9 cm) Long, Roboz

Supplier: Roboz
RS-8150
100189-384EA 65.08 USD InStock
100189-384
Fine Points, 3.5" (9 cm) Long, Roboz
Forceps
Splinter Forceps; Fine Points; 3.5" Length

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty