Would you like to log in again?
Forceps, 4" (10 cm) Long, 1×2 Teeth, Straight, 0.5 mm Tip, Roboz
Supplier: Roboz
RS-5102
100189-530EA 242.31 USD InStock
100189-530
Forceps, 4" (10 cm) Long, 1×2 Teeth, Straight, 0.5 mm Tip, Roboz
Forceps
Foerster Octagon Forceps; Straight; 1x2 Teeth; 0.5 mm Tip Width; 4" Length
The unique characteristic of the Foerster Forceps is the octagonal handle. The unique handle is designed to provide more "feel" and control at the tip. This handle also provides a better grip while wearing gloves. The tips are very fine and serrated and provide a sure, steady grip on tissue without trauma. The Foerster forceps are excellent for fine dissection of mice and dissection under a microscope.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty