Would you like to log in again?
Forceps, 4" (10 cm) Long, Serrated, Slight Curve, 0.8 mm Tip, Roboz
Supplier: Roboz
RS-5135
100189-532EA 98.31 USD InStock
100189-532
Forceps, 4" (10 cm) Long, Serrated, Slight Curve, 0.8 mm Tip, Roboz
Forceps
Micro Dissecting Forceps; Serrated; Slight Curve; 0.8 mm Tip Width; 4" Length
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty