Would you like to log in again?
Forceps, 7" (18 cm) Long, Straight With Suture Tying Platform, 0.7 mm Tip, Roboz
Supplier: Roboz
RS-5260
100189-608EA 420.92 USD InStock
100189-608
Forceps, 7" (18 cm) Long, Straight With Suture Tying Platform, 0.7 mm Tip, Roboz
Forceps
Rhoton Forceps; Micro Suturing With Tying Platform; 0.7 mm Tip Width; 7" Length.
Rhoton forceps can be used in a variety of surgical situations. The identifying feature of this instrument is its long, narrow tip which is helpful in procedures where you need to grasp and manipulate fine, small tissue in deep cavities.
Rotational movements can be performed by rolling the instrument gently between the fingers instead of twisting the wrist with this unusual round grip, which provides great control of the instrument and the tip. The box-style textured handle provides a "non slip" grip for even more control, particularly when wearing gloves. The Rhoton style is also manufactured in a spring action scissor (Roboz item number RS-5693). As with the Rhoton forceps, fine tissue in a deep cavity is easier to manipulate and cut with a long narrow tip.
Rhoton forceps can be used in a variety of surgical situations
The identifying feature of this instrument is its long, narrow tip which is helpful in procedures where you need to grasp and manipulate fine, small tissue in deep cavities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty