Would you like to log in again?
Forceps, 7" (18 cm) Long, Serrated Jaws, Sharp Scraper/Chisel End, Roboz
Supplier: Roboz
RS-5280
100189-674EA 105.23 USD InStock
100189-674
Forceps, 7" (18 cm) Long, Serrated Jaws, Sharp Scraper/Chisel End, Roboz
Forceps
Cushing Micro Dissecting Forceps; Scraper End; Serrated; 7" Length These special forceps have a chisel-like end with a very sharp edge that can be used for scraping.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty