Would you like to log in again?
Castroviejo Retractor 3.75" 15×5 mm, Roboz Surgical
Supplier: Roboz
RS-6570
100189-686EA 347.54 USD InStock
100189-686
Castroviejo Retractor 3.75" 15×5 mm, Roboz Surgical
Retractors
Castroviejo Retractor; Non-Magnetic; 35 mm Spread; 15 mm Blade Length; 5 mm Blade Depth; 3 3/4" Length
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty