Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Forceps, 4" (10 cm) Long, Angled Flat Jaws, Roboz

Supplier: Roboz
RS-8260
100189-820EA 56.77 USD InStock
100189-820
Forceps, 4" (10 cm) Long, Angled Flat Jaws, Roboz
Forceps
Kuehne Forceps; Flat Jaw; Angled; 4" Length

The Kuehne Micro Dissecting Forceps are reco mmended for picking up slides and cover glass. They have a unique thin flat, angled, bill-like tip that allows the fine glass to slip onto its tip easily.  They are also available in a straight version (RS-8261.)

The Kuehne Micro Dissecting Forceps are reco mmended for picking up slides and cover glass

They have a unique thin flat, angled, bill-like tip that allows the fine glass to slip onto its tip easily.  They are also available in a straight version (RS-8261.)

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty