Would you like to log in again?
Dumont #PP Heavy Duty Stainless Steel Forceps, Tip Size 0.10×0.06 mm, Biologie Tips, Roboz
Supplier: Roboz
RS-4950
100189-848EA 60.92 USD InStock
100189-848
Dumont #PP Heavy Duty Stainless Steel Forceps, Tip Size 0.10×0.06 mm, Biologie Tips, Roboz
Forceps
Dumont Tweezers; Pattern #PP; Heavy Duty; .10 x .06 mm Tip Size; Stainless Steel; 135 mm Length; Material: Inox; Biologie Tips
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty