Would you like to log in again?
Mayo-Hegar Needle Holder, Roboz
Supplier: Roboz
RS-7912L RS-7920 RS-7922 RS-7912 RS-7910
100189-816EA 303.23 USD InStock
100189-816 100189-900 100189-902 100189-814 100189-566
Mayo-Hegar Needle Holder, Roboz
Forceps
These Mayo-Hegar needle holders are recommended for 1–0 to 4–0 suture.
Caution: Note: Tungsten carbide inserts have 2600 teeth per square inch.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty