Would you like to log in again?
Bottle-top dispensers, Dispensette® S Organic
Supplier: Brand
Dispensette® S
4630341 4630160 4630170 4630230 4630360 4630140 4630330 4630151 4630331 4630161 4630351 4630240 4630241 4630131 4630361 4630171 4630340 4630141 4630350 4630130 4630231 4630150
10018-988EA 1325.02 USD InStock
10018-988 10018-984 10018-996 10018-986 10018-998 10019-020 10019-024 10019-002 10019-022 10019-000 10019-006 10020-974 10019-018 10019-004 10020-968 10019-008 10018-992 10018-982 10018-994 10020-970 10020-972 10018-990
Bottle-top dispensers, Dispensette® S Organic
Bottle Top Dispensers
Fixed volume or variable volume, analogue-adjustable or digital easy calibration.
  • With or without recirculation valve
  • Autoclavable at 121 °C
  • Three different models of volume adjustment available: fixed (fixed-volume for standard applications; simple calibration adjustment with supplied tool), analog (analog volume display, fast volume adjustment; simple calibration adjustment with supplied tool) and digital
  • Digital EASY CALIBRATION: Easy to read digital display; dispensing volume can be set accurately and reproducibly. Easy calibration technique for adjustment in seconds without tools; alteration of factory setting is automatically indicated
  • Easy to calibrate and adjust in order to comply with ISO 9001 and GLP guidelines; alteration of factory setting is automatically indicated
Dispensette® S Organic is ideal for dispensing organic solvents such as chlorinated and fluorinated hydrocarbons like trichlorotrifluoroethane and dichloromethane, or acids like concentrated HCl and HNO₃ (except for HF), as well as for trifluoroacetic acid (TFA), tetrahydrofurane (THF), and peroxides. For dispensing hydrofluoric acid (HF), we recommend the use of the Dispensette® S Trace Analysis bottle-top dispenser with platinum-iridium valve spring. Parts that are in contact with the liquid are made from borosilicate glass, ceramic, tantalum, ETFE, FEP, PFA, PTFE and PP (discharge tube safety screw cap). Colour code: yellow.
* Special fixed volumes: 0.5 to 100 ml (please state when ordering)!
Ordering information: Dispensette® S Organic supplied with DE-M marking, performance certificate, telescopic filling tube, recirculation tube (optional), mounting tool and PP adapters.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty