Would you like to log in again?
Halstead Mosquito Forceps, 5", Curved, Extra Delicate, 0.6 mm tips, Roboz
Supplier: Roboz
RS-7113
100190-000EA 130.15 USD InStock
100190-000
Halstead Mosquito Forceps, 5", Curved, Extra Delicate, 0.6 mm tips, Roboz
Forceps
Halstead Mosquito Forceps; Extra Delicate; Curved; 5" length, 0.6 mm tip diameter, 1.7 mm jaw width
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty