Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Intracardiac Needle Holder, 5" Extra Del Tungsten Carbide, Roboz

Supplier: Roboz
RS-7800
100190-004EA 343.38 USD InStock
100190-004
Intracardiac Needle Holder, 5" Extra Del Tungsten Carbide, Roboz
Forceps
Intracardiac Needle Holder; Extra Delicate; Carbide Jaws; 7" Length

Tapered, narrow jaws. Jaw width 1.3-1.5 mm. Note: Tungsten Carbide Inserts Have 4,000 Teeth Per Square Inch. 

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty