Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Forceps, 7" (18 cm) Long, Curved, 1×2 Teeth, Roboz

Supplier: Roboz
RS-5273
100190-044EA 113.54 USD InStock
100190-044
Forceps, 7" (18 cm) Long, Curved, 1×2 Teeth, Roboz
Forceps
Gerald Micro Dissecting Forceps; Curved, 1x2 Teeth; 7" Length

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty