Would you like to log in again?
Vet Scissors 6.5" Hvy Straight Blunt/Blunt Serr, Roboz Surgical
Supplier: Roboz
RS-6942
100190-194EA 193.85 USD InStock
100190-194
Vet Scissors 6.5" Hvy Straight Blunt/Blunt Serr, Roboz Surgical
Scissors
Veterinary Scissors; Heavy Pattern; Straight, Blunt-Blunt Serrated Blades; Blade Length 57 mm 6 1/2" Overall Length
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty