Would you like to log in again?
Extra Fine Micro Dissecting Scissors 4" Curved Sharp/Sharp, Roboz Surgical
Supplier: Roboz
RS-5883
100190-264EA 132.92 USD InStock
100190-264
Extra Fine Micro Dissecting Scissors 4" Curved Sharp/Sharp, Roboz Surgical
Scissors
Extra Fine Micro Dissecting Scissors; Curved; Sharp Points; 0.4 mm Combined Tip Width; 24 mm Blade Length; 4" Overall Length.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty