Would you like to log in again?
Doyen Abdominal Scissors 7" Curved, Roboz Surgical
Supplier: Roboz
RS-6931
100190-282EA 146.77 USD InStock
100190-282
Doyen Abdominal Scissors 7" Curved, Roboz Surgical
Scissors
Doyen Abdominal Scissors; Curved; 67 mm Blade Length ; 7" Overall Length
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty