Would you like to log in again?
Micro Dissecting Scissors 4.5" Straight Sharp/Sharp Left Handed, Roboz Surgical
Supplier: Roboz
RS-5912L
100190-316EA 168.92 USD InStock
100190-316
Micro Dissecting Scissors 4.5" Straight Sharp/Sharp Left Handed, Roboz Surgical
Scissors
Micro Dissecting Scissors; Left-Handed Version; Straight; Sharp Points; 24 mm Blade Length; 4 1/2" Overall Length
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty