Would you like to log in again?
Forceps, 5" (12.5 cm) Long, Curved With Suture Tying Platform, 0.3 mm Tip, Roboz
Supplier: Roboz
RS-5263
100190-388EA 411.23 USD InStock
100190-388
Forceps, 5" (12.5 cm) Long, Curved With Suture Tying Platform, 0.3 mm Tip, Roboz
Forceps
Rhoton Forceps; Micro Suturing With Tying Platform; Curved; 5" Length, 0.3 mm Tip Width.Rhoton forceps can be used in a variety of surgical situations
The feature of this instrument is its long, narrow tip which is helpful in procedures where you need to grasp and manipulate fine, small tissue in deep cavities. The unusual round, textured grip provides great control of the instrument and the tip
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty