Would you like to log in again?
Hartman Mosquito Forceps, 4", Curved, 0.8 mm tips, Roboz
Supplier: Roboz
RS-7101
100190-398EA 99.69 USD InStock
100190-398
Hartman Mosquito Forceps, 4", Curved, 0.8 mm tips, Roboz
Forceps
Hartman Mosquito Forceps; Curved; 4" Length, 0.8 mm tip diameter, 1.9 mm jaw width @ 5 mm from the tip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty