Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt(II) chloride hexahydrate ≥99.999% (Co basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Cobalt dichloride hexahydrateCobalt Chloride, 6-HydrateCobaltous chloride hexahydrateCobalt Chloride, Hexahydrate

93-2735-5G 93-2735-25G
100199-038EA 96.25 USD InStock
100199-038 100206-988
Cobalt(II) chloride hexahydrate ≥99.999% (Co basis), PURATREM
Cobalt (II) chloride hexahydrate


Formula: CoCl₂·6H₂O
MW: 237.93 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: 86 °C
Density: 1.924
Storage Temperature: Ambient
MDL Number: MFCD00149652
CAS Number: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 13,02462
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty