Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) oxide ≥97% (99+%-Cu)

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Copper monoxide granulatedCopper oxideCupric oxide

93-2943-100G 93-2943-500G
100197-802EA 66.52 USD InStock
100197-802 100205-492
Copper(II) oxide ≥97% (99+%-Cu)
Copper (II) oxide


Formula: CuO
MW: 79.55 g/mol
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 1362 °C
Density: 6.3…6.49
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9,III
Merck Index: 13,02674
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty