Would you like to log in again?
CANNON® CL Low Temperature Viscosity Standards, Cannon Instruments
Supplier: Cannon Instrument Company
CANNON®
9727-N16.128 9727-N31.004 9727-N04.128 9727-N32.128 9727-N32.016 9727-N24.128 9727-N16.032 9727-N32.5 9727-N14.016 9727-N20.016 9727-N20.128 9727-N14.128 9727-N14.500 9727-N35.128 9727-N37.032 9727-N31.128 9727-N31.500 9727-N02.5 9727-N07.128 9727-N08.5 9727-N28.004 9727-N37.128 9727-N12.500 9727-N36.128 9727-N18.128 9727-N14.128 9727-N04.128 9727-N33.128 9727-N11.016 9727-N22.016 9727-N24.032 9727-N38.032 9727-N13.004 9727-N11.500 9727-N30.016 9727-N11.128 9727-N02.500 9727-N18.5 9727-N13.016 9727-N30.032 9727-N35.032 9727-N24.128 9727-N06.016 9727-N31.128 9727-N22.128 9727-N16.004 9727-N18.128 9727-N31.016 9727-N37.032 9727-N08.128 9727-N20.5 9727-N33.032 9727-N10.500 9727-N35.500 9727-N28.5 9727-N33.5 9727-N37.016 9727-N10.016 9727-N37.016 9727-N09.004 9727-N02.128 9727-N06.032 9727-N22.500 9727-N20.004 9727-N14.016 9727-N26.032 9727-N13.128 9727-N36.5 9727-N09.5 9727-N11.016 9727-N14.5 9727-N12.128 9727-N10.128 9727-N37.500 9727-N32.016 9727-N16.500 9727-N13.032 9727-N12.016 9727-N09.032 9727-N08.500 9727-N26.500 9727-N09.128 9727-N04.016 9727-N14.032 9727-N33.016 9727-N07.032 9727-N09.016 9727-N09.500 9727-N34.500 9727-N33.128 9727-N18.500 9727-N30.5 9727-N34.128 9727-N07.128 9727-N24.016 9727-N18.032 9727-N11.004 9727-N06.016 9727-N30.128 9727-N02.032 9727-N30.016 9727-N26.016 9727-N09.128 9727-N16.032 9727-N13.5 9727-N38.5 9727-N26.128 9727-N18.032 9727-N35.128 9727-N22.032 9727-N12.032 9727-N33.016 9727-N31.5 9727-N36.500 9727-N28.500 9727-N04.004 9727-N24.500 9727-N18.016 9727-N04.032 9727-N31.032 9727-N35.016 9727-N20.500 9727-N13.032 9727-N08.016 9727-N33.500 9727-N36.032 9727-N36.128 9727-N06.128 9727-N08.032 9727-N08.128 9727-N30.032 9727-N24.5 9727-N13.016 9727-N36.016 9727-N38.016 9727-N02.128 9727-N34.032 9727-N26.016 9727-N06.128 9727-N34.128 9727-N28.016 9727-N11.032 9727-N08.016 9727-N24.032 9727-N22.016 9727-N37.5 9727-N34.016 9727-N22.128 9727-N04.500 9727-N20.032 9727-N04.032 9727-N10.016 9727-N16.016 9727-N35.004 9727-N30.500 9727-N07.5 9727-N32.032 9727-N28.128 9727-N28.128 9727-N02.016 9727-N38.128 9727-N02.016 9727-N36.016 9727-N07.016 9727-N20.128 9727-N34.032 9727-N07.016 9727-N07.500 9727-N10.032 9727-N35.032 9727-N16.5 9727-N13.500 9727-N08.032 9727-N37.004 9727-N16.016 9727-N31.032 9727-N10.032 9727-N36.032 9727-N33.032 9727-N11.032 9727-N35.5 9727-N12.016 9727-N33.004 9727-N38.032 9727-N38.500 9727-N31.016 9727-N04.5 9727-N30.128 9727-N16.128 9727-N02.032 9727-N20.016 9727-N06.500 9727-N14.032 9727-N04.016 9727-N28.016 9727-N10.5 9727-N28.032 9727-N28.032 9727-N26.032 9727-N07.032 9727-N34.5 9727-N32.032 9727-N12.128 9727-N07.004 9727-N24.004 9727-N06.032 9727-N12.032 9727-N09.032 9727-N20.032 9727-N22.032 9727-N11.128 9727-N12.5 9727-N35.016 9727-N18.016 9727-N22.5 9727-N24.016 9727-N37.128 9727-N11.5 9727-N26.5 9727-N26.128 9727-N10.128 9727-N32.500 9727-N13.128 9727-N34.016 9727-N09.016 9727-N32.128 9727-N06.5 9727-N02.004
10021-586EA 868.75 USD InStock
10021-586 10021-584 10021-582 10021-580 CA10021-516 CA10021-636 CA10021-998 CA10021-514 CA10021-634 CA10021-512 CA10021-996 CA10021-632 CA10021-510 10021-588 CA10021-630 CA10021-508 CA10021-628 CA10021-506 CA10021-640 10021-476 10021-598 10021-474 10021-596 10021-472 10021-594 10021-592 CA10021-648 CA10021-526 CA10021-646 CA10021-524 CA10021-644 CA10021-522 10021-478 CA10021-642 CA10021-520 CA10021-518 10021-590 10021-488 10021-486 10021-484 10021-482 CA10021-538 CA10021-658 CA10021-536 CA10021-656 CA10021-534 CA10021-654 CA10021-532 CA10021-652 10022-056 10022-058 CA10021-528 10021-480 CA10021-662 10021-498 CA10021-540 10021-496 CA10021-660 10021-494 10021-492 CA10021-546 CA10021-666 CA10021-544 CA10021-664 CA10021-542 10021-490 CA10021-552 10021-542 CA10021-672 10021-664 CA10021-550 10021-540 CA10021-670 10021-662 10021-660 CA10021-558 10021-548 CA10021-556 10021-546 CA10021-676 10021-668 CA10021-554 10021-544 CA10021-674 10021-666 CA10022-120 CA10022-000 10022-118 10022-114 CA10022-002 10022-116 CA10021-684 10021-674 CA10021-562 10021-554 10021-672 10021-552 CA10021-560 10021-670 CA10021-680 10021-550 CA10021-568 10021-678 CA10021-566 10021-558 10021-676 10021-556 CA10021-564 CA10022-116 CA10022-118 CA10022-114 CA10021-574 10021-564 10021-562 10021-684 10021-560 CA10021-570 10021-682 10021-680 CA10021-578 10021-568 10021-566 CA10021-576 CA10021-584 10021-576 CA10021-582 10021-574 10021-572 CA10021-580 10021-570 CA10021-588 CA10021-586 10021-578 10022-000 10022-002 10022-120 CA10021-596 10021-620 CA10021-474 CA10021-594 CA10021-592 10021-506 10021-628 10021-504 10021-626 10021-502 CA10021-478 10021-624 CA10021-598 10021-500 10021-622 CA10021-476 10021-618 10021-630 CA10021-486 10021-994 10021-510 CA10021-484 CA10021-480 10021-638 10021-518 10021-636 10021-516 10021-634 10021-514 10021-998 CA10021-488 10021-632 10021-512 10021-996 10021-508 10021-520 CA10021-496 10021-642 10021-640 CA10021-494 CA10021-492 CA10021-490 10021-528 10021-526 10021-648 10021-524 10021-646 10021-522 10021-644 CA10021-498 CA10022-064 CA10022-066 CA10022-060 CA10022-062 10021-652 10021-532 10021-650 10021-530 10021-658 10021-538 10021-656 10021-536 10021-654 10021-534 CA10022-058 CA10021-604 CA10021-602 10022-060 CA10021-600 10022-066 10022-062 10022-064 CA10021-614 10021-606 CA10021-612 10021-604 10021-602 CA10021-610 10021-600 CA10021-606 CA10021-626 10021-616 CA10021-504 10021-614 CA10021-624 CA10021-502 CA10021-622 10021-612 10021-610 CA10021-618 10021-608 CA10021-616
CANNON® CL Low Temperature Viscosity Standards, Cannon Instruments
Standards Viscometry Standards
CANNON® CL standards provide certified dynamic viscosity data at temperatures from –5 °C to –35 °C. These standards are used to calibrate the cold-cranking simulator for oil testing.
Certifications: Certified to ASTM D 5293 and SAE Specification J300.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty