Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium oxide ≥99%, Powder 1-5 µm

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Aluminium(III) oxideAlumina C

93-1339-250G 93-1339-1000G
100197-692EA 106.15 USD InStock
100197-692 100211-210
Aluminium oxide ≥99%, Powder 1-5 µm
Aluminium oxide


Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C
Melting Pt: 2045 °C
Density: 3,97
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty