Would you like to log in again?
PBS (Phosphate Buffered Saline), 1X, with EDTA, pH 7.4, Teknova
Supplier: Teknova
P0204 P0204 P0203 P0203
100216-522EA 128.43 USD InStock
100216-522 CA100216-522 100216-320 CA100216-320
PBS (Phosphate Buffered Saline), 1X, with EDTA, pH 7.4, Teknova
Buffers Biological Buffers
PBS buffer contains 137 mM sodium chloride, 7 mM sodium phosphate dibasic, 3 mM potassium chloride, 1.4 mM potassium phosphate monobasic, 4 mM EDTA pH 7.4
Suitable for cell culture and molecular biology applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty