Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Kanamycin sulfate solution 50 mg/mL

Supplier: TEKNOVA INC
Synonyms: Kanamycin MonosulphateKanamycin AKanamycin Monosulfate

K2125 K2125
100216-368EA 61.59 USD InStock
100216-368 CA100216-368
Kanamycin sulfate solution 50 mg/mL
Kanamycin sulphate
Kanamycin solution is used for the selection of resistant bacteria.


Formula: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
MDL Number: MFCD00070253
CAS Number: 25389-94-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty