Would you like to log in again?
LB Broth Bottled Media, Teknova
Supplier: Teknova
L8040 L8040 L8000 L8000 L8050 L8050
100216-388EA 59.38 USD InStock
CA100216-436 100216-388 CA100217-990 CA100216-388 100217-990 100216-436
LB Broth Bottled Media, Teknova
Media for Microbiology Prepared Media for Microbiology
Used For the maintenance and propagation of E.coli, and commonly used for the growth of cells for plasmid DNA production.
  • Available in multiple volumes
  • Sterile
Contains 1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride. Ship and store at room temperature.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty