Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloramphenicol USP

Supplier: TEKNOVA INC
Synonyms: Chloromycetin®(R,R)-ChloramphenicolTega-Cetin

C0325 C0325
100216-960EA 117.69 USD InStock
100216-960 CA100216-960
Chloramphenicol USP
Chloramphenicol
Chloramphenicol is a broad-spectrum antibiotic effective against most strains of Gram-positive and Gram-negative bacteria.

  • Inhibits protein synthesis by interacting with the 50S portion of the 70S ribosome
  • At higher concentrations can inhibit DNA synthesis

Bacteriostatic and inhibiting protein synthesis. Water-soluble, but not as stable in aqueous solutions and more susceptible to hydrolysis by UV light.


Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MW: 323.13 g/mol
MDL Number: MFCD00078159
CAS Number: 56-75-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty