Would you like to log in again?
Teknova LB Medium, Low Salt
Supplier: Teknova
L9195 L9190 L9199 L9199 L9195 L9190
100217-038EA 91.54 USD InStock
CA100219-284 CA100217-038 100217-038 100219-284 100217-714 CA100217-714
Teknova LB Medium, Low Salt
Media for Microbiology Dehydrated Media for Microbiology
Teknova's Dry Media is formulated to meet the needs of a molecular biologist
All Teknova Dry Media is made of the highest quality components and specifically tested for solubility, color, clarity,conductivity, pH, and growth assays. These convenient pre-mixes only require the addition of water and autoclaving. Lennox.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty