Would you like to log in again?
Super Broth, Teknova
Supplier: Teknova
S0150 S0145 S0140 S0135
100219-876EA 108.12 USD InStock
100219-876 100218-072 100219-646 100217-180
Super Broth, Teknova
Media for Microbiology Dehydrated Media for Microbiology
For use in propagation bacteriophage lambda.
  • Use 57 grams per liter to reconstitute
  • Ambient shipping conditions
  • Store at room temperature
Includes 3.2% tryptone, 2.0% yeast extract, and 0.5% sodium chloride, 0.99% Pottassium Phosphate, Dibasic.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty