Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 10 N

Supplier: TEKNOVA INC
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

N4710
100217-766EA 122.33 USD InStock
100217-766
Sodium hydroxide 10 N
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty