Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

G418 disulfate

Supplier: TEKNOVA INC
Synonyms: G-418 disulfateG-418 disulphateG418 disulfateG-418 sulphateG418 sulphateG-418 sulfateGeneticin®

G5005 G5001
100218-044EA 235.38 USD InStock
100218-044 100216-972
G418 disulfate
G418 disulphate
Geneticin is used as a selection marker in both prokaryotic and eukaryotic cell lines

  • Referred to as G418
  • Aminoglycosidic antibiotic
  • Similar to Neomycin, Kanamycin and Gentamycin

Geneticin, referred to as G418, is an aminoglycosidic antibiotic, that is similar to Neomycin, Kanamycin and Gentamycin. It is used as a selection marker in both prokaryotic and eukaryotic cell lines.


Formula: C₂₀H₄₄N₄O₁₈S₂
MW: 692.72 g/mol
MDL Number: MFCD00058314
CAS Number: 108321-42-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty