Would you like to log in again?
2X YT Broth, Teknova
Supplier: Teknova
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Y0205 Y0210 Y0220 Y0210 Y0205 Y0220 Y0215 Y0215
100218-396EA 2164.58 USD InStock
100218-396 CA100218-396 CA100219-840 CA100218-868 100219-840 100220-034 CA100220-034 100218-868
2X YT Broth, Teknova
Media for Microbiology Dehydrated Media for Microbiology
2X YT is a nutritionally rich medium for cultivating recombinant strains of Escherichia coli.
  • Use 31 grams per Liter to reconstitute media
  • pH 7.0
Ingredients (per Liter): 1.6% Tryptone (16.0g/L), 1.0% Yeast Extract (10.0g/L), and 0.5% Sodium Chloride (NaCl) (5.0g/L). Ambient shipping conditions. Store at room temperature.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty