Would you like to log in again?
MOPS Minimal Medium Kit, Teknova
Supplier: Teknova
M2106 M2106
100218-936EA 274.62 USD InStock
100218-936 CA100218-936
MOPS Minimal Medium Kit, Teknova
Media for Microbiology Prepared Media for Microbiology
Designed for the growth of E
MOPS mixture should be stored frozen. The potassium phosphate dibasic solution and 20% glucose solution should be stored at room temperature.
coli and other enterobacteria. Kit consists of 100mL 10X MOPS mixture (200059-678), 10mL 0.132M potassium phosphate diabase solution (200059-652), and 100mL 20% glucose solution (100216-728), and makes 5 liters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty