Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Kanamycin sulfate, white to off-white powder USP

Supplier: TEKNOVA INC
Synonyms: Kanamycin MonosulphateKanamycin AKanamycin Monosulfate

K2105 K2105
100218-998EA 52.31 USD InStock
100218-998 CA100218-998
Kanamycin sulfate, white to off-white powder USP
Kanamycin sulphate


Formula: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
MDL Number: MFCD00070253
CAS Number: 25389-94-0
Order Now
Specification Test Results
Potency 750 mg/ml
pH (1%, H₂O) 6.5 - 8.5
Moisture 4.0%
Ash 1.0%
Loss on Drying 4.0%
Residue on Ignition 1.0%
Chromatographic Purity Passes

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty