Would you like to log in again?
LB Broth, Teknova
Supplier: Teknova
L9135 L9145 L9130 L9140
100218-394EA 1778.46 USD InStock
100218-394 100218-046 100218-844 100219-108
LB Broth, Teknova
Media for Microbiology Dehydrated Media for Microbiology
For the growth of<italic> E
  • Use 25 grams per Liter to reconstitute
  • Ambient shipping conditions
  • Store at room temperature
Ingredients: 1.0% Tryptone (10.0 g/L), 0.5% Yeast Extract (5.0 g/L), and 1.0% NaCl (10.0 g/L).
coli</italic> and other enteric bacteria.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty