Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside), Ultrapure

Supplier: TEKNOVA INC
Synonyms: BCIG(2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

X1201 X1205 X1205 X1201
100219-246EA 91.54 USD InStock
CA100219-246 CA100218-934 100219-246 100218-934
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside), Ultrapure
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Used in conjunction with cloning systems that use b-galactosidase activity as an indicator of nonrecombination.

White powder


Formula: C₁₄H₁₅BrClNO₆
MW: 408.63 g/mol
MDL Number: MFCD00005666
CAS Number: 7240-90-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty