Would you like to log in again?
PBS (Phosphate Buffered Saline), 1X, 0.05% Tween®-20, Teknova
Supplier: Teknova
P0201
100219-788EA 158.15 USD InStock
100219-788
PBS (Phosphate Buffered Saline), 1X, 0.05% Tween®-20, Teknova
Buffers Biological Buffers
PBS/Tween for immunological and other applications.
Contains 137 mM sodium chloride, 2.7 mM potassium chloride, 4.3 mM disodium phosphate, 1.4 mM monopotassium phosphate and 0.05% (v/v) Tween-20 (polysorbate 20).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty