Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Citric acid solution 0.1 M, sterile

Supplier: TEKNOVA INC
Warning
Synonyms: Hydroxytricarballylic acid

C2440 C2440
100219-812EA 83.69 USD InStock
CA100219-812 100219-812
Citric acid solution 0.1 M, sterile
Citric acid


Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/50/46/725046.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty