Would you like to log in again?
DNA Suspension Buffer, Teknova
Supplier: Teknova
T0227 T0220 T0223 T0221 T0227 T0221
100219-272EA 74.89 USD InStock
100219-272 100216-886 CA100216-886 100217-094 100219-884 CA100217-094
DNA Suspension Buffer, Teknova
Buffers Cloning Buffers
RNAse, DNAse Tested (10mM Tris, 0.1mM EDTA)
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty