Would you like to log in again?
Ampicillin USP
Supplier: TEKNOVA INC
A9511 A9510 A9511 A9510
100219-890EA 91.54 USD InStock
CA100219-890 100219-890 CA100217-496 100217-496
Ampicillin USP
Ampicillin
Ampicillin is used as a selectable marker in many commonly used plasmids. A functional Ampicillin gene will confer resistance to cells containing the plasmid.
  • Inhibits cell wall synthesis by inhibiting formation of the peptidoglycan cross-linkages
  • Acts as a competitive inhibitor of transpeptidase
Inhibits bacterial cell wall synthesis by binding to one or more of the penicillin binding proteins (PBPs) that inhibit the final transpeptidation step of peptidoglycan synthesis in bacterial cell walls, thus inhibiting cell wall biosynthesis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty