Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Reagent Grade

Supplier: TEKNOVA INC
Synonyms: (p-tert-Octylphenoxy)polyethoxyethanolO-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanolOctoxynolPolyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether

T1100 T1100
100219-910EA 83.69 USD InStock
CA100219-910 100219-910
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether), Reagent Grade
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)


MDL Number: MFCD00132505
CAS Number: 9002-93-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty