Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium chloride 2 M in aqueous solution, sterile

Supplier: TEKNOVA INC
Synonyms: Potassiumchloride

P0330 P0330 P0335 P0335
100219-982EA 169.36 USD InStock
CA100219-982 CA100219-992 100219-982 100219-992
Potassium chloride 2 M in aqueous solution, sterile
Potassium chloride


Formula: KCl
MW: 74.55 g/mol
MDL Number: MFCD00011360
CAS Number: 7447-40-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty