Would you like to log in again?
Screw Cap Sample Tubes, Globe Scientific
Supplier: Globe Scientific
6144
10022-134CS 698.71 USD InStock
10022-134
Screw Cap Sample Tubes, Globe Scientific
Tubes Test Tubes
These tubes and caps have been designed for the storage and transportation of biological material. They are manufactured from non-toxic polypropylene and offer strength and clarity.
  • Polypropylene screw caps provide same coefficient of expansion as the tubes
  • Leakproof seal even at changing temperatures
The vial has external threads, which provide a smooth and uniform inner surface and reduces the risk of contamination.
The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty