Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glass Capillary Tubes, Globe Scientific

Supplier: Globe Scientific
51608 51602 51613
10022-154BX 91.45 USD InStock
10022-154 10022-156 10022-152
Glass Capillary Tubes, Globe Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Glass capillary tubes are designed for both safe blood collection as well as accurate micro-hematocrit determinations

  • IØ: 1.15 mm (±0.05 mm)
  • OØ: 1.5 mm (±0.05 mm)
  • Length: 75 mm (±0.05 mm)
  • Volume: 75 µl
  • Spin Rate: 14500 rpm for 5 Minutes

Tubes are available plain or with an anticoagulant (sodium or ammonium heparin) to prevent blood from clotting. All tubes feature color-coded ends to easily identify the plain and heparin treated tubes.

Precision molded, tubes provide excellent sample retention during centrifugation. In addition, the tubes fill rapidly without affecting specimen integrity.

Packaging: Tubes are packaged 100 per vial, 10 vials per box (1000 tubes per box).

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty