Would you like to log in again?
Corning® Gosselin™ Storage Bottles, Square, HDPE, Corning
Supplier: Corning
Corning®
LC2500A-07 LC250-06 LC150A-05 LC2000-03 LC500-11 LC1000-13
10799-438CS 210.26 USD InStock
10799-438 10022-310 10799-436 10022-306 10022-308 10022-312
Corning® Gosselin™ Storage Bottles, Square, HDPE, Corning
Bottles Plastic Bottles
Space saving bottles made of HDPE, translucent with HDPE or LDPE screw cap.
  • Good chemical resistance, good temperature resistance
  • Sterile oder packed under aseptic conditions
  • Excellent shock resistance
Ideal for liquid, solid and powder storage. Good level of tightness according to the closure system used and application.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty